Learn More
Biolegend Alexa Fluor 647 anti-mouse CD68
Supplier:  Biolegend 137004
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD68 [FA-11]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: ICFC, FC; Size: 100 ug
Catalog No. 50-164-938