Learn More
Biolegend Alexa Fluor 647 anti-mouse CD80
Supplier:  Biolegend 104717
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC, IHC-F; Size: 25 ug
Catalog No. 50-162-978