Learn More
Biolegend Alexa Fluor 647 anti-mouse CD9
Supplier:  Biolegend 124810
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD9 [MZ3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0626