Learn More
Biolegend Alexa Fluor 647 anti-mouse CXCL9 (MIG)
Supplier:  Biolegend 515606
Encompass_Preferred
Alexa Fluor 647 anti-mouse CXCL9 (MIG) [MIG-2F5.5]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-170-197