Learn More
Biolegend Alexa Fluor 647 anti-mouse/human CD324 (E-Cadherin)
Supplier:  Biolegend 147307
Encompass_Preferred
Alexa Fluor 647 anti-mouse/human CD324 (E-Cadherin) [DECMA-1]; Isotype: Rat IgG1, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Dog, Swine; Apps: FC, ICC, IHC-F; Size: 25 ug
Catalog No. 50-850-401