Learn More
Biolegend Alexa Fluor 647 anti-mouse Ikaros
Supplier:  Biolegend 653306
Encompass_Preferred
Alexa Fluor 647 anti-mouse Ikaros [2A9/Ikaros]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-208-7041