Learn More
Biolegend Alexa Fluor 647 anti-mouse IL-17F
Supplier:  Biolegend 517104
Encompass_Preferred
Alexa Fluor 647 anti-mouse IL-17F [8F5.1A9]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-170-223