Learn More
Biolegend Alexa Fluor 647 anti-mouse IL-4
Supplier:  Biolegend 504110
Encompass_Preferred
Alexa Fluor 647 anti-mouse IL-4 [11B11]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-169-832