Learn More
Biolegend Alexa Fluor 647 anti-mouse RAE-1γ
Supplier:  Biolegend 130110
Encompass_Preferred
Alexa Fluor 647 anti-mouse RAE-1γ [CX1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-553