Learn More
Biolegend Alexa Fluor 647 anti-mouse TCR β chain
Supplier:  Biolegend 109217
Encompass_Preferred
Alexa Fluor 647 anti-mouse TCR β chain [H57-597]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F; Size: 25 ug
Catalog No. 50-163-351