Learn More
Biolegend Alexa Fluor 647 anti-Myelin Basic Protein
Supplier:  Biolegend 808408
Encompass_Preferred
Alexa Fluor 647 anti-Myelin Basic Protein [SMI 99]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P; Size: 100 ug
Catalog No. 50-207-4008