Learn More
Biolegend Alexa Fluor 647 anti-OCT4 (OCT3)
Supplier:  Biolegend 653710
Encompass_Preferred
Alexa Fluor 647 anti-OCT4 (OCT3) [3A2A20]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: ICC, ICFC; Size: 100 ug
Catalog No. 50-208-7051