Learn More
Biolegend Alexa Fluor 647 anti-Tau, 269-281
Supplier:  Biolegend 850603
Encompass_Preferred
Alexa Fluor 647 anti-Tau, 269-281 [A16040D]; Isotype: Rat IgG2b, κ; Reactivity: Human, Mouse; Apps: IHC-P; Size: 25 ug
Catalog No. 50-207-4606