Learn More
Biolegend Alexa Fluor 647 anti-ZAP70 Phospho (Tyr292)
Supplier:  Biolegend 693807
Encompass_Preferred
Alexa Fluor 647 anti-ZAP70 Phospho (Tyr292) [A16038B]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-208-7640