Learn More
Biolegend Alexa Fluor 647 Mouse IgG2b, κ isotype Ctrl
Supplier:  Biolegend 400330
Encompass_Preferred
Alexa Fluor 647 Mouse IgG2b, κ isotype Ctrl [MPC-11]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse ; Apps: FC, ICFC; Size: 100 ug
Catalog No. NC9664367