Learn More
Biolegend Alexa Fluor 647 Mouse IgG3, κ isotype Ctrl
Supplier:  Biolegend 401322
Encompass_Preferred
Alexa Fluor 647 Mouse IgG3, κ isotype Ctrl [MG3-35]; Isotype: Mouse IgG3, κ; Apps: FC; Size: 100 ug
Catalog No. 50-169-041