Learn More
ABclonal Technology FOXF1 Rabbit mAb
SDP
Catalog No. 5000135513
Encompass_Preferred
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined, however, it may play a role in the regulation of pulmonary genes as well as embryonic development.
Catalog No. 50-001-35513 Supplier ABclonal Technology Supplier No. A6513500UL
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined, however, it may play a role in the regulation of pulmonary genes as well as embryonic development.