Learn More
Apexbio Technology LLC A66 1166227-08-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061668
Encompass_Preferred
A66 (CAS 1166227-08-2) is a selective inhibitor of the p110 catalytic subunit of class I phosphatidylinositol-4 5-bisphosphate 3-kinase (PI3K) with an IC50 of 32 nM This compound specifically decreases phosphorylation of Akt/PKB and downstream S6K at Thr389 in various cell models including PIK3CA-mutant H1047R cell lines C2C12 myoblasts and 3T3-L1 adipocytes In animal studies A66 reduces tumor growth in xenograft models and improves insulin sensitivity under chronic metabolic stress highlighting its utility in research on PI3K signaling in cancer and metabolic disease
Catalog No. 50-000-61668 Supplier Apexbio Technology LLC Supplier No. A83545.1
 Restricted Item
Add to Cart
Add to Cart
A66 (CAS 1166227-08-2) is a selective inhibitor of the p110 catalytic subunit of class I phosphatidylinositol-4 5-bisphosphate 3-kinase (PI3K) with an IC50 of 32 nM This compound specifically decreases phosphorylation of Akt/PKB and downstream S6K at Thr389 in various cell models including PIK3CA-mutant H1047R cell lines C2C12 myoblasts and 3T3-L1 adipocytes In animal studies A66 reduces tumor growth in xenograft models and improves insulin sensitivity under chronic metabolic stress highlighting its utility in research on PI3K signaling in cancer and metabolic disease