Learn More
Selleck Chemical LLC A66 5mg 1166227-08-2
SDP
Supplier:  Selleck Chemical LLC S26365MG
Encompass_Preferred
A66 is a potent and specific p110? inhibitor with IC50 of 32 nM in a cell-free assay, >100 fold selectivity for p110? over other class-I PI3K isoforms. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7987