Learn More
Selleck Chemical LLC NU7441 (KU-57788) 5mg 503468-95-9
SDP
Supplier:  Selleck Chemical LLC S26385MG
Encompass_Preferred
NU7441 (KU-57788) is a highly potent and selective DNA-PK inhibitor with IC50 of 14 nM and also inhibits mTOR and PI3K with IC50 of 1.7 ?M and 5 ?M in cell-free assays, respectively. It reduces the frequency of NHEJ while increasing the rate of HDR following Cas9-mediated DNA cleavage. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0513442