Learn More
MSC North Disposable Gloves, Tri-Polymer, Small, 5.9 mil, Powder-Free
Catalog No. 5000508075
Encompass
North PowerCoat disposable gloves are designed for chemical and liquid protection. They are made from a tri-polymer blend of latex, nitrile, and neoprene, offering durability and comfort. These powder-free gloves are ambidextrous and non-sterile, suitable for various general purpose and chemical-resistant applications.
  • Offers excellent chemical and liquid protection.
  • Designed for limited use and are disposable.
  • Are powder free.
  • Made from a blend of latex, nitrile, and neoprene for durability.
  • Feature a smooth texture and rolled cuff for comfort.
  • Are ambidextrous and non-sterile.
  • Have an extended cuff type.
Catalog No. 50-005-08075 Supplier MSC Supplier No. 59704783
North PowerCoat disposable gloves are designed for chemical and liquid protection. They are made from a tri-polymer blend of latex, nitrile, and neoprene, offering durability and comfort. These powder-free gloves are ambidextrous and non-sterile, suitable for various general purpose and chemical-resistant applications.
  • Offers excellent chemical and liquid protection.
  • Designed for limited use and are disposable.
  • Are powder free.
  • Made from a blend of latex, nitrile, and neoprene for durability.
  • Feature a smooth texture and rolled cuff for comfort.
  • Are ambidextrous and non-sterile.
  • Have an extended cuff type.