Learn More
Biolegend Alexa Fluor 700 anti-human CD172a/b (SIRPα/β)
Supplier:  Biolegend 323815
Encompass_Preferred
Alexa Fluor 700 anti-human CD172a/b (SIRPα/β) [SE5A5]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-207-5617