Learn More
Biolegend Alexa Fluor 700 anti-human CD271 (NGFR)
Catalog No. 502076754
Encompass_Preferred
Alexa Fluor 700 anti-human CD271 (NGFR) [ME20.4]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-207-6754 Supplier Biolegend Supplier No. 345118
Add to Cart
Add to Cart
Alexa Fluor 700 anti-human CD271 (NGFR) [ME20.4]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests