Learn More
Biolegend Alexa Fluor 700 anti-human CD335 (NKp46)
Supplier:  Biolegend 331932
Encompass_Preferred
Alexa Fluor 700 anti-human CD335 (NKp46) [9E2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-6073