Learn More
Biolegend Alexa Fluor 700 anti-human CD4
Supplier:  Biolegend 317425
Encompass_Preferred
Alexa Fluor 700 anti-human CD4 [OKT4]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 25 ug
Catalog No. 50-605-264