Learn More
Biolegend Alexa Fluor 700 anti-human CD45RA
Supplier:  Biolegend 304120
Encompass_Preferred
Alexa Fluor 700 anti-human CD45RA [HI100]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 ug
Catalog No. 50-166-083