Learn More
Biolegend Alexa Fluor 700 anti-human CD90 (Thy1)
Supplier:  Biolegend 328120
Encompass_Preferred
Alexa Fluor 700 anti-human CD90 (Thy1) [5E10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Pigtailed Macaque, Rhesus, Swine (Pig, Porcine); Apps: FC; Size: 100 ug
Catalog No. 50-167-425