Learn More
Biolegend Alexa Fluor 700 anti-human TCR α/β
Supplier:  Biolegend 306729
Encompass_Preferred
Alexa Fluor 700 anti-human TCR α/β [IP26]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-3321