Learn More
Biolegend Alexa Fluor 700 anti-human TNF-α
Supplier:  Biolegend 502928
Encompass_Preferred
Alexa Fluor 700 anti-human TNF-α [MAb11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cat (Feline), Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Swine (Pig, Porcine), Pigtailed Macaque, Sooty Mangabey; Apps: ICFC, FC; Size: 100 tests
Catalog No. 50-604-738