Learn More
Biolegend Alexa Fluor 700 anti-human CD38
Supplier:  Biolegend 397206
Encompass_Preferred
Alexa Fluor 700 anti-human CD38 [S17015F]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-9296