Learn More
Biolegend Alexa Fluor 700 anti-human β2-microglobulin
Supplier:  Biolegend 395708
Encompass_Preferred
Alexa Fluor 700 anti-human β2-microglobulin [A17082A]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-9193