Learn More
Biolegend Alexa Fluor 700 anti-mouse CD45.1
Supplier:  Biolegend 110724
Encompass_Preferred
Alexa Fluor 700 anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC9908067