Learn More
Biolegend Alexa Fluor 700 anti-mouse CD48
Supplier:  Biolegend 103426
Encompass_Preferred
Alexa Fluor 700 anti-mouse CD48 [HM48-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-851