Learn More
Biolegend Alexa Fluor 700 anti-mouse CD62L
Supplier:  Biolegend 104426
Encompass_Preferred
Alexa Fluor 700 anti-mouse CD62L [MEL-14]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-927