Learn More
Biolegend Alexa Fluor 700 anti-mouse/human CD44
Supplier:  Biolegend 103025
Encompass_Preferred
Alexa Fluor 700 anti-mouse/human CD44 [IM7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Squirrel Monkey, Horse (Equine), Cattle (Bovine, Cow), Swine (Pig, Porcine), Dog (Canine), Cat (Feline); Apps: FC; Size: 25 ug
Catalog No. 50-162-764