Learn More
Biolegend Alexa Fluor 700 anti-mouse Ly-6A/E (Sca-1)
Supplier:  Biolegend 108142
Encompass_Preferred
Alexa Fluor 700 anti-mouse Ly-6A/E (Sca-1) [D7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0015