Learn More
Aviva Systems Biology Corp Endov Antibody - middle region (ARP55675_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55675_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against A730011L01Rik. It was validated on Western Blot. . Alternate names: Endov, A730011L01RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-286-064