Learn More
Enzo Life Sciences A771726 (5mg). CAS: 108605-62-5
SDP
Supplier:  Enzo Life Sciences ALX430096M005
Encompass_Preferred
Physiologically active metabolite of the immunosuppressive drug leflunomide (ALX-430-095). Inhibits the activity of dihydrorotate dehydrogenase and of protein tyrosine kinases. Blocks TNF-mediated NF-κB activation in a dose- and time-dependent manner. Also inhibits the activity of cyclooxygenase-2 (COX-2) in vitro and in vivo. Alternative name: N-(4-Trifluoromethylphenyl)-2-cyano-3-hydroxycrotonamide, 2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]-2-butenamide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: +4°C.
Catalog No. 50-200-9199