Learn More
Abcam A771726 (Teriflunomide), Active metabolite of leflunomide. Dihydroorotate dehydrogenase inhibitor., 25MG
Catalog No. 502706582
Encompass_Preferred
MW 270.21 Da, Purity >98%. Active metabolite of leflunomide. Inhibits dihydroorotate dehydrogenase. Shows antiproliferative effects. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6582 Supplier Abcam Supplier No. AB14148025MG
MW 270.21 Da, Purity >98%. Active metabolite of leflunomide. Inhibits dihydroorotate dehydrogenase. Shows antiproliferative effects. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement