Learn More
ABclonal Technology PKC delta Rabbit mAb
SDP
Supplier:  ABclonal Technology A7778500UL
Encompass_Preferred
The protein encoded by this gene is a member of the protein kinase C family of serine- and threonine-specific protein kinases. The encoded protein is activated by diacylglycerol and is both a tumor suppressor and a positive regulator of cell cycle progression. Also, this protein can positively or negatively regulate apoptosis. Defects in this gene are a cause of autoimmune lymphoproliferative syndrome.
Catalog No. 50-001-34857