Learn More
ABclonal Technology PSD95 Rabbit PolymAb
SDP
Supplier:  ABclonal Technology A7889PM1000UL
Encompass_Preferred
This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) family. It heteromultimerizes with another MAGUK protein, DLG2, and is recruited into NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-34771