Learn More
ATCC A7r5; Aorta; Rat (Rattus norvegicus)
Catalog No. 502384354
Encompass
A7r5 is a fibroblast-like cell isolated from the aortic thoracic smooth muscle of an embryonic rat. This cell line was deposited by W Carlisle and can be used in cardiovascular disease research.
Catalog No. 50-238-4354 Supplier ATCC Supplier No. CRL1444
A7r5 is a fibroblast-like cell isolated from the aortic thoracic smooth muscle of an embryonic rat. This cell line was deposited by W Carlisle and can be used in cardiovascular disease research.