Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ A8 Agar for Mycoplasma and Ureaplasma
SDP

Catalog No. 23001776 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Species:
Ureaplasma parvum, Mycoplasma hominis, Staphylococcus epidermidis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species
23-001-776 Ureaplasma parvum, Mycoplasma hominis, Staphylococcus epidermidis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-776 Supplier Hardy Diagnostics™ Supplier No. G02
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the selective isolation of Mycoplasma hominis and Ureaplasma urealyticum.

  • A8 Agar is highly nutritious due to peptones supplemented with yeast extract and inactivated horse serum present in the medium
    • The yeast extract provides diphosphopyridine nucleotides and the serum provides cholesterol and a source of protein
  • Amphotericin B and penicillin G are added to inhibit faster growing contaminants
  • Optically clear polystyrene for distortion free viewing
  • Stacking rings guard against slippage when moving stacked plates

Specifications

Appearance Slightly Opalescent
Color Whitish to Light Amber
Content And Storage 2°C to 8°C
Additive Amphotericin B, Penicillin G
Detects Mycoplasma hominis, Ureaplasma urealyticum
Format Plate
Product Type Agar Plate
Target Organism Class Mycobacteria
Fill Volume 11 mL
For Use With (Application) Selective Isolation
Incubation Conditions 35°C
Incubation Time 24 hr., 72 to 96 hr.
Material Polystyrene
No. per Pack 10
Packaging Type Sleeve, Plates, Breathable Bag
Quantity 10/Pk.
Dimensions (Dia. x H) 60 x 15 mm
Diameter (Metric) 100 mm
Species Ureaplasma parvum, Mycoplasma hominis, Staphylococcus epidermidis
Protein Source Peptones, Yeast Extract, Horse Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.