Learn More
Cayman Chemical A83016F 2.5mg
SDP
Catalog No. 502986938
Encompass_Preferred
A DNA alkylating agent active metabolite of dacarbazine (DTIC) active degradation product of temozolomide cytotoxic to L-cells and decreases thymidine and uridine uptake by 55 and 65 respectively at 1 mM cytotoxic to TLX5 murine lymphoma cells induces formation of mammary adenofibromas in rats when administered at a cumulative dose of 890 mg per animal over 14 weeks
Catalog No. 50-298-6938 Supplier Cayman Chemical Supplier No. 232402.5 MG
A DNA alkylating agent active metabolite of dacarbazine (DTIC) active degradation product of temozolomide cytotoxic to L-cells and decreases thymidine and uridine uptake by 55 and 65 respectively at 1 mM cytotoxic to TLX5 murine lymphoma cells induces formation of mammary adenofibromas in rats when administered at a cumulative dose of 890 mg per animal over 14 weeks