Learn More
ABclonal Technology p150 CAF1 Rabbit mAb
SDP
Catalog No. 5000134866
Encompass_Preferred
Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B, MIM 601245), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro (Kaufman et al., 1995 [PubMed 7600578]).
Catalog No. 50-001-34866 Supplier ABclonal Technology Supplier No. A87251000UL
Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B, MIM 601245), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro (Kaufman et al., 1995 [PubMed 7600578]).