Learn More
ABclonal Technology MSH2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A87401000UL
Encompass_Preferred
This locus is frequently mutated in hereditary nonpolyposis colon cancer (HNPCC). When cloned, it was discovered to be a human homolog of the E. coli mismatch repair gene mutS, consistent with the characteristic alterations in microsatellite sequences (RER+ phenotype) found in HNPCC. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-37160