Learn More
ABclonal Technology MYL2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A87421000UL
Encompass_Preferred
This gene encodes a major sarcomeric protein in mammalian striated muscle. The encoded protein plays a role in embryonic heart muscle structure and function, while phosphorylation of the encoded protein is involved in cardiac myosin cycling kinetics, torsion and function in adults. Mutations in this gene are associated with hypertrophic cardiomyopathy 10 and infant-onset myopathy.
Catalog No. 50-001-34743