Learn More
ABclonal Technology PRKRA/PACT Rabbit mAb
SDP
Catalog No. 5000134898
Encompass_Preferred
This gene encodes a protein kinase activated by double-stranded RNA which mediates the effects of interferon in response to viral infection. Mutations in this gene have been associated with dystonia. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-34898 Supplier ABclonal Technology Supplier No. A8779500UL
This gene encodes a protein kinase activated by double-stranded RNA which mediates the effects of interferon in response to viral infection. Mutations in this gene have been associated with dystonia. Alternative splicing results in multiple transcript variants.