Learn More
Cayman Chemical A8SGLP-1 (trifluoroacetate salt), Purity: ≥98%, CAS: 753024-08-7, 5 mg
SDP
Catalog No. 502944496
Encompass_Preferred
A peptide agonist of GLP-1R and a derivative of GLP-1 (7-36) amide; induces cAMP accumulation in RIN T3 cells (EC50 = 15 nM); reduces blood levels of glucose in the OGTT, serum levels of Il-1β, Il-6, and Tnf-α, and fat mass in diabetic db/db mice in lyophilized L. lactis, p.o.
Catalog No. 50-294-4496 Supplier Cayman Chemical Supplier No. 412645 MG
A peptide agonist of GLP-1R and a derivative of GLP-1 (7-36) amide; induces cAMP accumulation in RIN T3 cells (EC50 = 15 nM); reduces blood levels of glucose in the OGTT, serum levels of Il-1β, Il-6, and Tnf-α, and fat mass in diabetic db/db mice in lyophilized L. lactis, p.o.