Learn More
ATCC A9; Connective Tissue; Mouse (Mus musculus)
Catalog No. 502383238
Encompass
A9 (APRT and HPRT negative derivative of Strain L) is a cell line isolated from wild-type Strain L cells that's sensitive to HAT selection medium and deficient in adenosine phosphoribosyl transferase (APRT) and hypoxanthine phosphoribosyl transferase (HPRT).
Catalog No. 50-238-3238 Supplier ATCC Supplier No. CCL1.4
A9 (APRT and HPRT negative derivative of Strain L) is a cell line isolated from wild-type Strain L cells that's sensitive to HAT selection medium and deficient in adenosine phosphoribosyl transferase (APRT) and hypoxanthine phosphoribosyl transferase (HPRT).